Learn More
Grainger Blucher® Joint Clamp, 316 Stainless Steel, 2 in., 1 ea.
Catalog No. 502753095
Encompass_Preferred
This stainless steel joint clamp is designed for secure pipe and hose connections, featuring a durable 316 stainless steel construction finished with pickling and passivation for enhanced corrosion resistance. It includes an EPDM sealing ring to ensure a tight, leak-proof seal in demanding industrial environments.
  • Constructed from 316 stainless steel for optimal durability
  • Features pickling and passivation finish process
  • Equipped with an EPDM sealing ring
  • Suitable for 2 in. pipe sizes
  • Certified to the Uniform Plumbing Code (ASME A112.6.3)
  • Compliant with NSF/ANSI/3-A 14159-1 hygiene standards for food processing equipment
Catalog No. 50-275-3095 Supplier Grainger Supplier No. 14P012
This stainless steel joint clamp is designed for secure pipe and hose connections, featuring a durable 316 stainless steel construction finished with pickling and passivation for enhanced corrosion resistance. It includes an EPDM sealing ring to ensure a tight, leak-proof seal in demanding industrial environments.
  • Constructed from 316 stainless steel for optimal durability
  • Features pickling and passivation finish process
  • Equipped with an EPDM sealing ring
  • Suitable for 2 in. pipe sizes
  • Certified to the Uniform Plumbing Code (ASME A112.6.3)
  • Compliant with NSF/ANSI/3-A 14159-1 hygiene standards for food processing equipment