Learn More
Grainger Joint Clamp 3 In 316SS
Catalog No. 502753096
Encompass_Preferred
The BLUCHER Joint Clamp is a stainless steel clamp used to connect drainage pipes and fittings. It has a working pressure of up to 10 bar and temperature range of -20°C to +95°C. The clamp is available in sizes ranging from 40 to 200mm diameter pipes. It provides a secure, watertight connection and can accommodate angular deflection up to 4°. The clamp has a V-shaped cross section and uses a single bolt to tighten it around the pipe joint. It is made from AISI 304 stainless steel for corrosion resistance. The clamp complies with EN 1253 standards.
Catalog No. 50-275-3096 Supplier Grainger Supplier No. 14P013
The BLUCHER Joint Clamp is a stainless steel clamp used to connect drainage pipes and fittings. It has a working pressure of up to 10 bar and temperature range of -20°C to +95°C. The clamp is available in sizes ranging from 40 to 200mm diameter pipes. It provides a secure, watertight connection and can accommodate angular deflection up to 4°. The clamp has a V-shaped cross section and uses a single bolt to tighten it around the pipe joint. It is made from AISI 304 stainless steel for corrosion resistance. The clamp complies with EN 1253 standards.