Learn More
Grainger Joint Clamp 4 In 316SS
Catalog No. 502753050
Encompass_Preferred
The Blucher Joint Clamp is a stainless steel pipe clamp designed for joining pipes and tubes. It has a working pressure of 232 psi and temperature range of -4 to 392 degrees Fahrenheit. This clamp features a V-shaped cross section with extended lips to ensure maximum rigidity. It is made from 304 stainless steel for corrosion resistance. The clamp has a bolt hole diameter of 0.4 inches and comes in sizes ranging from 1/2 inch to 8 inch. Overall, this is a heavy duty stainless pipe clamp well-suited for joining and repairing pipes in industrial applications.
Catalog No. 50-275-3050 Supplier Grainger Supplier No. 14P014
The Blucher Joint Clamp is a stainless steel pipe clamp designed for joining pipes and tubes. It has a working pressure of 232 psi and temperature range of -4 to 392 degrees Fahrenheit. This clamp features a V-shaped cross section with extended lips to ensure maximum rigidity. It is made from 304 stainless steel for corrosion resistance. The clamp has a bolt hole diameter of 0.4 inches and comes in sizes ranging from 1/2 inch to 8 inch. Overall, this is a heavy duty stainless pipe clamp well-suited for joining and repairing pipes in industrial applications.