Learn More
Sigma Aldrich Fine Chemicals Biosciences Joint clip standard taper
Catalog No. 502906160
Encompass
Color coded rigid polyethylene joint clips fit standard taper joints. Will not scratch glassware or corrode withstands temperatures up to 120 C.
Catalog No. 50-290-6160 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z75931750EA
Color coded rigid polyethylene joint clips fit standard taper joints. Will not scratch glassware or corrode withstands temperatures up to 120 C.