Learn More
Ace Glass, Inc. Joint clips, 19/22, Keck; type, POM acetal resin, not for positive pressure use, blue, pk/10
SDP
Catalog No. NC0096925
Encompass
19/22 blue Keck type joint clips made from POM acetal resin. Clips will not scratch the glass, and are resistant to concentrated alkalies and dilute acids. Useful temp range: -40 to 140 C. Not for pressure applications. Pk/10.
Catalog No. NC0096925 Supplier Ace Glass, Inc. Supplier No. 759819
19/22 blue Keck type joint clips made from POM acetal resin. Clips will not scratch the glass, and are resistant to concentrated alkalies and dilute acids. Useful temp range: -40 to 140 C. Not for pressure applications. Pk/10.