Learn More
Grainger Joint Knife Straight SS 4 W
Catalog No. 502767731
Encompass_Preferred
The Hyde Joint Knife Blade Material Stainless Steel is a utility knife blade made of stainless steel. It has a total length of 2-1/2 inches and a cutting edge of 2 inches. The blade thickness is 0.025 inches and it fits most standard utility knives that use trapezoidal blades. This durable stainless steel blade is ideal for cutting cardboard, plastic, carpet, and other common materials. It features a sharp edge and pointed tip to allow for precise cuts. The trapezoidal shank securely fastens into the knife handle. This is a high quality utility knife blade for everyday use.
Catalog No. 50-276-7731 Supplier Grainger Supplier No. 16W166
The Hyde Joint Knife Blade Material Stainless Steel is a utility knife blade made of stainless steel. It has a total length of 2-1/2 inches and a cutting edge of 2 inches. The blade thickness is 0.025 inches and it fits most standard utility knives that use trapezoidal blades. This durable stainless steel blade is ideal for cutting cardboard, plastic, carpet, and other common materials. It features a sharp edge and pointed tip to allow for precise cuts. The trapezoidal shank securely fastens into the knife handle. This is a high quality utility knife blade for everyday use.