Learn More
Grainger Joint Knife Straight SS 5 W
Catalog No. 502766280
Encompass_Preferred
The Hyde Joint Knife Blade Material Stainless Steel is a utility knife blade made of stainless steel. It features a trapezoidal shape with two sharp edges and measures 2-3/4" long x 7/8" wide x 0.050" thick. The blade has a hardness of Rc54-56 and fits most standard utility knife handles that accept trapezoidal blades. It can be used for a variety of cutting tasks including opening boxes, cutting rope and fabric. This durable stainless steel blade stays sharp longer than carbon steel. It is sold individually.
Catalog No. 50-276-6280 Supplier Grainger Supplier No. 16W167
The Hyde Joint Knife Blade Material Stainless Steel is a utility knife blade made of stainless steel. It features a trapezoidal shape with two sharp edges and measures 2-3/4" long x 7/8" wide x 0.050" thick. The blade has a hardness of Rc54-56 and fits most standard utility knife handles that accept trapezoidal blades. It can be used for a variety of cutting tasks including opening boxes, cutting rope and fabric. This durable stainless steel blade stays sharp longer than carbon steel. It is sold individually.