Learn More
Chemglass Life Sciences O-Ring Joint, 25mm ID, 30mm Tube OD , O-Ring Size: 217, Clamp Size: 35
SDP
Catalog No. 50873705
Encompass_Preferred
O-Ring Joint, 25mm ID, 30mm Tube OD , O-Ring Size: 217, Clamp Size: 35. Joint has an o-ring groove to accept the listed o-ring size. Provides for a grease-free, vacuum seal when used with CG-150 clamps. Each joint is supplied with a Viton o-ring of the listed size.
Catalog No. 50-873-705 Supplier Chemglass Life Sciences Supplier No. CG12406
O-Ring Joint, 25mm ID, 30mm Tube OD , O-Ring Size: 217, Clamp Size: 35. Joint has an o-ring groove to accept the listed o-ring size. Provides for a grease-free, vacuum seal when used with CG-150 clamps. Each joint is supplied with a Viton o-ring of the listed size.