Learn More
Chemglass Life Sciences O-Ring Joint, 30mm ID, 35mm Tube OD , O-Ring Size: 223, Clamp Size: 50
SDP
Catalog No. 50873706
Encompass_Preferred
O-Ring Joint, 30mm ID, 35mm Tube OD , O-Ring Size: 223, Clamp Size: 50. Joint has an o-ring groove to accept the listed o-ring size. Provides for a grease-free, vacuum seal when used with CG-150 clamps. Each joint is supplied with a Viton o-ring of the listed size.
Catalog No. 50-873-706 Supplier Chemglass Life Sciences Supplier No. CG12407
O-Ring Joint, 30mm ID, 35mm Tube OD , O-Ring Size: 223, Clamp Size: 50. Joint has an o-ring groove to accept the listed o-ring size. Provides for a grease-free, vacuum seal when used with CG-150 clamps. Each joint is supplied with a Viton o-ring of the listed size.