Learn More
Chemglass Life Sciences #20 Quartz O-Ring Joint, 23.0mm Tube O.D., Clamp Size/35, 214 O-Ring Size
SDP
Catalog No. 501532057
Encompass_Preferred
#20 Quartz O-Ring Joint, 23.0mm Tube O.D., Clamp Size/35, 214 O-Ring Size. Clear fused quartz o-ring joint. Mating surface is formed so that quartz joints can be connected directly to CG-124 borosilicate o-ring joints. For clamps see CG-150.
Catalog No. 50-153-2057 Supplier Chemglass Life Sciences Supplier No. CGQ013506
#20 Quartz O-Ring Joint, 23.0mm Tube O.D., Clamp Size/35, 214 O-Ring Size. Clear fused quartz o-ring joint. Mating surface is formed so that quartz joints can be connected directly to CG-124 borosilicate o-ring joints. For clamps see CG-150.