Learn More
Cole-Parmer Scienceware 20320-1660 PTFE Joint Stirrer Bearing for Standard Taper Size 24/40; 1/Pk
Catalog No. NC1634031 Shop All Cole-Parmer Products
Encompass_Preferred
Use with 10-mm OD stirring rods. Use O-ring for fine adjustment of rod tension. Threaded PP locking compression cap. Removable bushing fits inside adapter (extra bushing included).
Catalog No. NC1634031 Supplier Cole-Parmer Supplier No. 0613985
Use with 10-mm OD stirring rods. Use O-ring for fine adjustment of rod tension. Threaded PP locking compression cap. Removable bushing fits inside adapter (extra bushing included).