Learn More
Chemglass Life Sciences 45/50 Outer Joint, 16in OAL, on 57mm OD x 2.4mm W tubing
SDP
Catalog No. 501531003
Encompass_Preferred
45/50 Outer Joint, 16in OAL, on 57mm OD x 2.4mm W tubing. These joints offer the glassblower the opportunity to construct apparatus made of heavier tubing or without unnecessary seals.
Catalog No. 50-153-1003 Supplier Chemglass Life Sciences Supplier No. CG10949
45/50 Outer Joint, 16in OAL, on 57mm OD x 2.4mm W tubing. These joints offer the glassblower the opportunity to construct apparatus made of heavier tubing or without unnecessary seals.