Learn More
MSC Jomac Heat Resistant Gloves, Heavyweight Terrycloth, 450 F, 1 Dz.
Catalog No. 5000673917
Encompass
Jomac 644HRL heavy weight heat resistant gloves are designed to provide contact heat protection in high-temperature applications.
  • Heavyweight terrycloth material
  • Provides contact heat resistance up to 450°F
  • Safety cuff with a 2-1/2 in. cuff, loop-out
  • Lined and uncoated design
  • Ambidextrous design for extended wear
  • Full-finger style for complete hand protection
  • Available in various sizes
  • Brown and white color
Catalog No. 50-006-73917 Supplier MSC Supplier No. 43650738
Jomac 644HRL heavy weight heat resistant gloves are designed to provide contact heat protection in high-temperature applications.
  • Heavyweight terrycloth material
  • Provides contact heat resistance up to 450°F
  • Safety cuff with a 2-1/2 in. cuff, loop-out
  • Lined and uncoated design
  • Ambidextrous design for extended wear
  • Full-finger style for complete hand protection
  • Available in various sizes
  • Brown and white color