Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Jomac™ Trapper's Gloves

Catalog No. 19048727 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Overall length: 31 in., Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-048-727 Overall length: 31 in., Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-048-727 Supplier Wells Lamont™ Supplier No. S22120BLK

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Black
Material PVC, Nylon
Size Small
Liner Material Interlock Knit
Lining Lined
Length (English) 31 in.
Length (Metric) 78.4 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.