Learn More
MSC Jonard Tools Force Gauge, Push-Pull, 0-72 oz, 0-2025 g
Catalog No. 5000528403
Encompass
This push-pull force gauge is economically constructed with a high impact polystyrene casing. It accurately measures push-pull forces, with applications including testing belt tension. The gauge offers dual scales in grams and ounces, making it ideal for field service and maintenance-free operation.
  • Economically constructed with high impact polystyrene casing.
  • Accurately measures push-pull forces.
  • Can be used to test belt tension in ATMs.
  • Features dual graduated scales: grams and ounces.
  • Ideal for field service and maintenance-free operation.
  • Precision springs are individually calibrated and checked.
  • Scales maintain full accuracy (± 1 graduation).
  • Maximum capacity: 72 oz (2025 g).
Catalog No. 50-005-28403 Supplier MSC Supplier No. 40598070
This push-pull force gauge is economically constructed with a high impact polystyrene casing. It accurately measures push-pull forces, with applications including testing belt tension. The gauge offers dual scales in grams and ounces, making it ideal for field service and maintenance-free operation.
  • Economically constructed with high impact polystyrene casing.
  • Accurately measures push-pull forces.
  • Can be used to test belt tension in ATMs.
  • Features dual graduated scales: grams and ounces.
  • Ideal for field service and maintenance-free operation.
  • Precision springs are individually calibrated and checked.
  • Scales maintain full accuracy (± 1 graduation).
  • Maximum capacity: 72 oz (2025 g).