Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Jones and Foster Salt Mixture

Catalog No. MP290211002 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 kg
10 kg
50 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP290211002 2 kg
MP290211010 10 kg
MP290211050 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP290211002 Supplier MP Biomedicals™ Supplier No. 0290211002

Salt mixture for research animal diet.

  • Salt Mixture
  • For research animal diet
  • Powder
TRUSTED_SUSTAINABILITY

Specifications

Format Powder
For Use With (Application) Research animal diet
Quantity 2 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.