Learn More
Med Vet International Jor-Vet Cuffed Endotracheal Tube 4.5mm, J0149YC
Catalog No. 5000583691
Encompass_Preferred
Softens at body temperature, reducing postoperative irritation. Blue-tinted PVC tube complete with connector.Available in 0.5mm increments for 3.5mm to 11mm
Catalog No. 50-005-83691 Supplier Med Vet International Supplier No. JOR149YC
Softens at body temperature, reducing postoperative irritation. Blue-tinted PVC tube complete with connector.Available in 0.5mm increments for 3.5mm to 11mm