Learn More
Med Vet International Jor-Vet Cuffed Endotracheal Tube, J0149XC, 4.0mm, Each
Catalog No. 5000583633
Encompass_Preferred
Softens at body temperature, reducing postoperative irritation. Blue-tinted PVC tube complete with connector.Available in 0.5mm increments for 3.5mm to 11mm
Catalog No. 50-005-83633 Supplier Med Vet International Supplier No. JOR149XC
Softens at body temperature, reducing postoperative irritation. Blue-tinted PVC tube complete with connector.Available in 0.5mm increments for 3.5mm to 11mm