Learn More
Med Vet International Jorgensen Osteotome, 10mm, J0228A
Catalog No. 5000585390
Encompass_Preferred
Shorter length for finer control, suggestedfor femoral head removal. German-made. Stainlesssteel. 13.5cm (5").
Catalog No. 50-005-85390 Supplier Med Vet International Supplier No. JOR228A
Shorter length for finer control, suggestedfor femoral head removal. German-made. Stainlesssteel. 13.5cm (5").