Learn More
Med Vet International Jorgensen Yankauer Suction Handle, Metal, J0489M, Each
Catalog No. 5000581834
Encompass_Preferred
Small removable tip has six vertical holes for suction. Fits standard suction hose. 22cm L.
Catalog No. 50-005-81834 Supplier Med Vet International Supplier No. JOR489M
Small removable tip has six vertical holes for suction. Fits standard suction hose. 22cm L.