Learn More
Med Vet International Jorvet 14g x 9cm Super Core Biopsy Needle, J0528
Catalog No. 5000581984
Encompass_Preferred
Jorvet 14g x 9cm super core biopsy needle, is sterile and not autoclavable. It has a spring activated single handle operation, is lightweight, has a echogenic tip for ultrasound visualization, and a 20mm specimen notch. Each needle is marked cm graduations.
Catalog No. 50-005-81984 Supplier Med Vet International Supplier No. JOR528
Jorvet 14g x 9cm super core biopsy needle, is sterile and not autoclavable. It has a spring activated single handle operation, is lightweight, has a echogenic tip for ultrasound visualization, and a 20mm specimen notch. Each needle is marked cm graduations.