Learn More
Med Vet International Jorvet 16g x 15cm Super Core Biopsy Needle, J0528B
Catalog No. 5000581557
Encompass_Preferred
Jorvet 16g x 15cm super core biopsy needle, is sterile and not autoclavable. It has a spring activated single handle operation, is lightweight, has a echogenic tip for ultrasound visualization, and a 20mm specimen notch. Each needle is marked cm graduations.
Catalog No. 50-005-81557 Supplier Med Vet International Supplier No. JOR528B
Jorvet 16g x 15cm super core biopsy needle, is sterile and not autoclavable. It has a spring activated single handle operation, is lightweight, has a echogenic tip for ultrasound visualization, and a 20mm specimen notch. Each needle is marked cm graduations.