Learn More
Med Vet International Jorvet 18g x 15cm Chiba Style Biopsy Needle, J0527A
Catalog No. 5000581983
Encompass_Preferred
Jorvet 18g x 15cm Chiba style biopsy needle, is a general purpose aspiration needle. It has a shor bevel to remove samples from controlled areas. It attaches to standard syringes. This product is sterile and not autoclavable.
Catalog No. 50-005-81983 Supplier Med Vet International Supplier No. JOR527A
Jorvet 18g x 15cm Chiba style biopsy needle, is a general purpose aspiration needle. It has a shor bevel to remove samples from controlled areas. It attaches to standard syringes. This product is sterile and not autoclavable.