Learn More
Med Vet International Jorvet 20g x 15cm Chiba Ultrasound Biopsy Needle, J0527EC
Catalog No. 5000581803
Encompass_Preferred
Jorvet 20g x 15cm Chiba ultrasound biopsy needle, is a general purpose aspiration needle. It has a shor bevel to remove samples from controlled areas. It attaches to standard syringes. This product is sterile and not autoclavable.
Catalog No. 50-005-81803 Supplier Med Vet International Supplier No. JOR527EC
Jorvet 20g x 15cm Chiba ultrasound biopsy needle, is a general purpose aspiration needle. It has a shor bevel to remove samples from controlled areas. It attaches to standard syringes. This product is sterile and not autoclavable.