Learn More
Med Vet International Jorvet 20g x 9cm Chiba Ultrasound Biopsy Needle, J0527EB
Catalog No. 5000582299
Encompass_Preferred
Jorvet 20g x 9cm Chiba ultrasound biopsy needle, is a general purpose aspiration needle. It has a shor bevel to remove samples from controlled areas. It attaches to standard syringes. This product is sterile and not autoclavable.
Catalog No. 50-005-82299 Supplier Med Vet International Supplier No. JOR527EB
Jorvet 20g x 9cm Chiba ultrasound biopsy needle, is a general purpose aspiration needle. It has a shor bevel to remove samples from controlled areas. It attaches to standard syringes. This product is sterile and not autoclavable.