Learn More
Med Vet International Jorvet 20g x 9cm Franseen Style Biopsy Needle, J0526B
Catalog No. 5000582364
Encompass_Preferred
Jorvet 20g x 9cm. Franseen style biopsy needle, is used to core boipsy specimens for histology and impression smears. This needle attaches to standard syringes. The cannula point design is titable for cutting difficult lesions. These needles are sterile and not autoclavable.
Catalog No. 50-005-82364 Supplier Med Vet International Supplier No. JOR526B
Jorvet 20g x 9cm. Franseen style biopsy needle, is used to core boipsy specimens for histology and impression smears. This needle attaches to standard syringes. The cannula point design is titable for cutting difficult lesions. These needles are sterile and not autoclavable.