Learn More
Med Vet International Jorvet 20g x 9cm Spinal Needle, J0529B
Catalog No. 5000581986
Encompass_Preferred
Jorvet 20g x 9cm spinal needle, is used as in gerneral purpose aspiration biopsies and drainage procedures. Each needle has a matched ground bevel that eliminates the possibility of tissue coring. They attach to a standard luer fitting and is sterile and not autoclavable.
Catalog No. 50-005-81986 Supplier Med Vet International Supplier No. JOR529B
Jorvet 20g x 9cm spinal needle, is used as in gerneral purpose aspiration biopsies and drainage procedures. Each needle has a matched ground bevel that eliminates the possibility of tissue coring. They attach to a standard luer fitting and is sterile and not autoclavable.