Learn More
Med Vet International Jorvet 22g x 15cm Franseen Style Biopsy Needle, J0526C
Catalog No. 5000582297
Encompass_Preferred
Jorvet 22g x 9cm. Franseen style biopsy needle, is used to core boipsy specimens for histology and impression smears. This needle attaches to standard syringes. The cannula point design is titable for cutting difficult lesions. These needles are sterile and not autoclavable.
Catalog No. 50-005-82297 Supplier Med Vet International Supplier No. JOR526C
Jorvet 22g x 9cm. Franseen style biopsy needle, is used to core boipsy specimens for histology and impression smears. This needle attaches to standard syringes. The cannula point design is titable for cutting difficult lesions. These needles are sterile and not autoclavable.