Learn More
Med Vet International Jorvet 22g x 3 1/2in. Spinal Needle, J0529H
Catalog No. 5000582000
Encompass_Preferred
Jorvet 22g x 3 1/2in. spinal needle, is used as in gerneral purpose aspiration biopsies and drainage procedures. Each needle has a matched ground bevel that eliminates the possibility of tissue coring. They attach to a standard luer fitting and is sterile and not autoclavable.
Catalog No. 50-005-82000 Supplier Med Vet International Supplier No. JOR529H
Jorvet 22g x 3 1/2in. spinal needle, is used as in gerneral purpose aspiration biopsies and drainage procedures. Each needle has a matched ground bevel that eliminates the possibility of tissue coring. They attach to a standard luer fitting and is sterile and not autoclavable.