Learn More
Med Vet International Jorvet 22g x 6in. Spinal Needle, J0529K
Catalog No. 5000581785
Encompass_Preferred
Jorvet 22g x 6in. spinal needle, is used as in gerneral purpose aspiration biopsies and drainage procedures. Each needle has a matched ground bevel that eliminates the possibility of tissue coring. They attach to a standard luer fitting and is sterile and not autoclavable.
Catalog No. 50-005-81785 Supplier Med Vet International Supplier No. JOR529K
Jorvet 22g x 6in. spinal needle, is used as in gerneral purpose aspiration biopsies and drainage procedures. Each needle has a matched ground bevel that eliminates the possibility of tissue coring. They attach to a standard luer fitting and is sterile and not autoclavable.