Learn More
Med Vet International Jorvet 4in. Dose Adaptor, J0206
Catalog No. 5000583123
Encompass_Preferred
Jorvet 4in. Dose adaptor that fits on all standard luer lock and luer tip syringes.
Catalog No. 50-005-83123 Supplier Med Vet International Supplier No. JOR206
Jorvet 4in. Dose adaptor that fits on all standard luer lock and luer tip syringes.