Learn More
Med Vet International Jorvet 50cc Hauptner Syringe, J0031C
Catalog No. 5000583097
Encompass_Preferred
50cc, adjusts to 1, 2, 3, 4, and 5cc doses.
Catalog No. 50-005-83097 Supplier Med Vet International Supplier No. JOR31C
50cc, adjusts to 1, 2, 3, 4, and 5cc doses.