Learn More
Med Vet International Jorvet 5cc Vetamatic Syringe, J0053V94
Catalog No. 5000581778
Encompass_Preferred
A handy, sturdy automatic multidose syringe. Plastic valve and washer reduces swelling problems.NOTE: Spare Parts Ordering: Parts 13, 1494 and 1694 as shown on parts diagram are not available individually. They can be purchased only as a repair kit: J-53vd 2894
Catalog No. 50-005-81778 Supplier Med Vet International Supplier No. JOR53V94
A handy, sturdy automatic multidose syringe. Plastic valve and washer reduces swelling problems.NOTE: Spare Parts Ordering: Parts 13, 1494 and 1694 as shown on parts diagram are not available individually. They can be purchased only as a repair kit: J-53vd 2894