Learn More
Med Vet International Jorvet Artificial Insemination Kit, for Small Animals, J0102
Catalog No. 5000585724
Encompass_Preferred
A complete kit for the veterinarian for the collection of semen and insemination of the female. Features: Latex collection cone Four vaginal pipettes 12cc syringe Small plastic cap for end of syringe Cone holder Direction sheet J-Jelly lubricating gel 8oz flask.
Catalog No. 50-005-85724 Supplier Med Vet International Supplier No. JOR102
A complete kit for the veterinarian for the collection of semen and insemination of the female. Features: Latex collection cone Four vaginal pipettes 12cc syringe Small plastic cap for end of syringe Cone holder Direction sheet J-Jelly lubricating gel 8oz flask.