Learn More
Med Vet International Jorvet Bone Marrow Needle, 13g x 4in, J0116S, Each
Catalog No. 5000581220
Encompass_Preferred
Jorvet bone marrow biopsy aspiration needle has a diamater of 13g and a length of 4in. These needles allow for an actual biopsy versus the aspriation other needles provide. A special reamer is included to allow retrieval of actual tissue samples. The tip of the needle has a double side diamond. This product is sterile and not autoclavable. NY, CT, IL, NJ, NH, DE and MN restrict the sale of syringes and needles to licensed professionals and entities. By proceeding to purchase through our website, you acknowledge that you are over 18 years of age.
Catalog No. 50-005-81220 Supplier Med Vet International Supplier No. JOR116S
Jorvet bone marrow biopsy aspiration needle has a diamater of 13g and a length of 4in. These needles allow for an actual biopsy versus the aspriation other needles provide. A special reamer is included to allow retrieval of actual tissue samples. The tip of the needle has a double side diamond. This product is sterile and not autoclavable. NY, CT, IL, NJ, NH, DE and MN restrict the sale of syringes and needles to licensed professionals and entities. By proceeding to purchase through our website, you acknowledge that you are over 18 years of age.