Learn More
Med Vet International Jorvet Bone Marrow Needle, 16g, J0116T, Each
Catalog No. 5000580955
Encompass_Preferred
The Jorvet 16g bone marrow needle has a regular tip, a large hand grip for increased torque, and an adjustable depth stop from 3/8in. to 1-7/8in shaft. This needle has a luer lock syringe attachment and is similar to a Jamshidi style. This produc can also be used for intraosseous fluid administration and is sterile and not autoclavable. NY, CT, IL, NJ, NH, DE and MN restrict the sale of syringes and needles to licensed professionals and entities. By proceeding to purchase through our website, you acknowledge that you are over 18 years of age.
Catalog No. 50-005-80955 Supplier Med Vet International Supplier No. JOR116T
The Jorvet 16g bone marrow needle has a regular tip, a large hand grip for increased torque, and an adjustable depth stop from 3/8in. to 1-7/8in shaft. This needle has a luer lock syringe attachment and is similar to a Jamshidi style. This produc can also be used for intraosseous fluid administration and is sterile and not autoclavable. NY, CT, IL, NJ, NH, DE and MN restrict the sale of syringes and needles to licensed professionals and entities. By proceeding to purchase through our website, you acknowledge that you are over 18 years of age.