Learn More
Med Vet International Jorvet Calf Gun, 2-3/8in. Head, 1in. Diameter, J0185
Catalog No. 5000585359
Encompass_Preferred
The Jorvet Balling gun is a simple deviced used for oral administration of capsules and boluses to animals. It has a 2-3/8in. Head and a 1in. Diameter.
Catalog No. 50-005-85359 Supplier Med Vet International Supplier No. JOR185
The Jorvet Balling gun is a simple deviced used for oral administration of capsules and boluses to animals. It has a 2-3/8in. Head and a 1in. Diameter.