Learn More
Med Vet International JorVet Central Venous Catheter Kit, 7fr x 20cm, Triple Lumen, J1045A, 1 Kit
Catalog No. 5000583407
Encompass_Preferred
These long-term catheters have been the standard of critical care in human medicine for many years. The catheters are placed in the jugular vein using a technique called Seldinger guide wire system. Non-reactive polyurethane catheter can be left in place for extended use i.e. 3-4 weeks Single, double or even triple lumen catheters of various lengths. Allows administration concurrently of different IV medications. Polyurethane radiopaque catheter with clamp Introducer needle Vessel dilator Guide wire Injection caps 5mL syringe Illustrated directions
Catalog No. 50-005-83407 Supplier Med Vet International Supplier No. JOR1045A
These long-term catheters have been the standard of critical care in human medicine for many years. The catheters are placed in the jugular vein using a technique called Seldinger guide wire system. Non-reactive polyurethane catheter can be left in place for extended use i.e. 3-4 weeks Single, double or even triple lumen catheters of various lengths. Allows administration concurrently of different IV medications. Polyurethane radiopaque catheter with clamp Introducer needle Vessel dilator Guide wire Injection caps 5mL syringe Illustrated directions