Learn More
Med Vet International JorVet Clinch Cutter, For Opening Clinches on Hooves, 6in, J1024C, Each
Catalog No. 5000585776
Encompass_Preferred
Clinch cutter. 6" double ended tool for opening the clinches to reduce hoof wall damage during shoe removal. Beveled on inside edge for closer cutting. Stainless. Other end is pointed for pritched.
Catalog No. 50-005-85776 Supplier Med Vet International Supplier No. JOR1024C
Clinch cutter. 6" double ended tool for opening the clinches to reduce hoof wall damage during shoe removal. Beveled on inside edge for closer cutting. Stainless. Other end is pointed for pritched.