Learn More
Med Vet International Jorvet Deluxe IV Stand with 4 hooks, J0266
Catalog No. 5000585288
Encompass_Preferred
24" diameter, five-leg aluminum base is more stable than the economy model. Brushed chrome, dual caster, and four prong hooks. Adjustable height: 54"-84".
Catalog No. 50-005-85288 Supplier Med Vet International Supplier No. JOR266
24" diameter, five-leg aluminum base is more stable than the economy model. Brushed chrome, dual caster, and four prong hooks. Adjustable height: 54"-84".