Learn More
Med Vet International Jorvet Detorsion Bar, 31.5in, J0121, Each
Catalog No. 5000584137
Encompass_Preferred
Used in bovine uterine torsion. Stainless steel. (chain notincluded) 311/2"L.
Catalog No. 50-005-84137 Supplier Med Vet International Supplier No. JOR121
Used in bovine uterine torsion. Stainless steel. (chain notincluded) 311/2"L.