Learn More
Med Vet International JorVet Duo Drape, 45cm x 75cm, Fenestration and Adhesive Patch, 10/Pack, J1456A
Catalog No. 5000583287
Encompass_Preferred
JorVet introduces a new bonded two-layer surgical drape! Outer layer is an absorbent layer of hydrophilic polypropylene. It will readily absorb body fluids such as blood. Inner layers is fluid repellant polyethylene that will prevent bodily fluids from leaking through the surgery field onto the patient. Adhesive patch on inner layer will adhere to patient preventing drape from moving during surgery • Fenestrations of various sizes • Inexpensive: Check out the cost! All come in 10 packs, individually wrapped Size: 45cm (18") x 75cm (30") Fenestration Size: 5cm (2") wide x 15cm (6") long
Catalog No. 50-005-83287 Supplier Med Vet International Supplier No. JOR1456A
JorVet introduces a new bonded two-layer surgical drape! Outer layer is an absorbent layer of hydrophilic polypropylene. It will readily absorb body fluids such as blood. Inner layers is fluid repellant polyethylene that will prevent bodily fluids from leaking through the surgery field onto the patient. Adhesive patch on inner layer will adhere to patient preventing drape from moving during surgery • Fenestrations of various sizes • Inexpensive: Check out the cost! All come in 10 packs, individually wrapped Size: 45cm (18") x 75cm (30") Fenestration Size: 5cm (2") wide x 15cm (6") long