Learn More
Med Vet International JorVet Endotracheal Tube, Uncuffed, J0149X, 4.0mm, Each
Catalog No. 5000583311
Encompass_Preferred
Un-Cuffed Endotracheal Tube. Softens at body temperature, reducing postoperative irritation. Blue-tinted PVC tube complete with connector.
Catalog No. 50-005-83311 Supplier Med Vet International Supplier No. JOR149X
Un-Cuffed Endotracheal Tube. Softens at body temperature, reducing postoperative irritation. Blue-tinted PVC tube complete with connector.