Learn More
Med Vet International Jorvet Equine Leg Wrap, Large Bulk Roll, 16" x 10 yds., J0849B
Catalog No. 5000582181
Encompass_Preferred
Made of soft quality bulk cotton which is covered on both sides with a permeable thin layer of nonwoven material. The wrap acts much like a diaper in that the wound exudate is lifted and absorbed into the thick middle cotton layer. A dark mark mid-line runs full length to give proper orientation during application. A wide variety of materials and sizes are now available:
Catalog No. 50-005-82181 Supplier Med Vet International Supplier No. JOR849B
Made of soft quality bulk cotton which is covered on both sides with a permeable thin layer of nonwoven material. The wrap acts much like a diaper in that the wound exudate is lifted and absorbed into the thick middle cotton layer. A dark mark mid-line runs full length to give proper orientation during application. A wide variety of materials and sizes are now available: