Learn More
Med Vet International Jorvet Extended Use Polyurethane Catheter, J0458B, 16g x 5.25in, Each
Catalog No. 5000581676
Encompass_Preferred
Most traditional jugular catheters are teflon. They can create a thrombosis and crack if left in place for over 48 hours. The development of new materials allows large-bore catheters that can safely be left in-dwelling for up to 10 days. The JorVet Extended Use Catheter softens at body temperature. Many of the common catheter-related problems, such as phlebitis, can be minimized with these catheters.14g and 16g polyurethane catheter over needleNon-thrombogenic and non-crackingCan be safely left in the vein for up to 10 daysThis same catheter is used in 24 out of 27 U.S.veterinary teaching hospitalsSuture wings with holes or suture groove attachment
Catalog No. 50-005-81676 Supplier Med Vet International Supplier No. JOR458B
Most traditional jugular catheters are teflon. They can create a thrombosis and crack if left in place for over 48 hours. The development of new materials allows large-bore catheters that can safely be left in-dwelling for up to 10 days. The JorVet Extended Use Catheter softens at body temperature. Many of the common catheter-related problems, such as phlebitis, can be minimized with these catheters.14g and 16g polyurethane catheter over needleNon-thrombogenic and non-crackingCan be safely left in the vein for up to 10 daysThis same catheter is used in 24 out of 27 U.S.veterinary teaching hospitalsSuture wings with holes or suture groove attachment