Learn More
Med Vet International Jorvet Feeding Needle, 16g x 2in, Curved, J0194CCE, Each
Catalog No. 5000582794
Encompass_Preferred
Jorvet 16g x 2in. Curved animal feeding needle has a luer lock hub and has a stainless steel tube and nickel-plated silver ball on the end.
Catalog No. 50-005-82794 Supplier Med Vet International Supplier No. JOR194CCE
Jorvet 16g x 2in. Curved animal feeding needle has a luer lock hub and has a stainless steel tube and nickel-plated silver ball on the end.