Learn More
Med Vet International JorVet Fiberglass Casting Tape, 2" x 4 Yards, J0574A, 5/Case
Catalog No. 5000581449
Encompass_Preferred
A woven fiberglass mesh that is impregnated with polyurethane. It is activated by submersion in water. Many competing tapes are only 3 yards long!Lightweight yet durableWaterproofEasily shapedPackaged in foil pouch
Catalog No. 50-005-81449 Supplier Med Vet International Supplier No. JOR574A
A woven fiberglass mesh that is impregnated with polyurethane. It is activated by submersion in water. Many competing tapes are only 3 yards long!Lightweight yet durableWaterproofEasily shapedPackaged in foil pouch