Learn More
Med Vet International Jorvet FUS Needle, 21g x 1/2in. Olive Tipped, J0153C
Catalog No. 5000583151
Encompass_Preferred
Jorvet 21g x 1/2in. olive tipped FUS needles are perfect for flushing of the small male feline urethra in cases of obstructive feline lower urinary tract disease (FLUTD). The blunt tip prevents trauma to the snesitive urethral lining. Rather than forcing struvite plugs back into the bladder with saline. it is suggested that J0153W Walpole's solution is infused to dissovle away any crystal formation. Then a J0153 series Jackson catheter is inserted to ensure urethral patency. This needle has a luer mount that fits any syringe and is stainless steel, and autoclavable.
Catalog No. 50-005-83151 Supplier Med Vet International Supplier No. JOR153C
Jorvet 21g x 1/2in. olive tipped FUS needles are perfect for flushing of the small male feline urethra in cases of obstructive feline lower urinary tract disease (FLUTD). The blunt tip prevents trauma to the snesitive urethral lining. Rather than forcing struvite plugs back into the bladder with saline. it is suggested that J0153W Walpole's solution is infused to dissovle away any crystal formation. Then a J0153 series Jackson catheter is inserted to ensure urethral patency. This needle has a luer mount that fits any syringe and is stainless steel, and autoclavable.