Learn More
Med Vet International Jorvet Hwato Accupuncture Needle, 0.25mm x 25mm, J1121D
Catalog No. 5000580939
Encompass_Preferred
Jorvet Hwato stainless steel Chinese style acupuncture needles have a sprial wound copper handle. Each needle is individually sealed in its own insertion tube. These needles measure: Needle Diamater: 0.25mm Japanese/Chinese Gauage: 5/#32 Needle Length: 1in.
Catalog No. 50-005-80939 Supplier Med Vet International Supplier No. JOR1121D
Jorvet Hwato stainless steel Chinese style acupuncture needles have a sprial wound copper handle. Each needle is individually sealed in its own insertion tube. These needles measure: Needle Diamater: 0.25mm Japanese/Chinese Gauage: 5/#32 Needle Length: 1in.