Learn More
Med Vet International Jorvet Hypodermic Needle, 16g x 1in., J0174CC, 12/box
Catalog No. 5000583126
Encompass_Preferred
Jorvet 16g x 1in. Hypodermic needle is made of stainless steel, has a round hub, and is packaged in a cardboard box or clear plastic case. This product contains 12 needles.
Catalog No. 50-005-83126 Supplier Med Vet International Supplier No. JOR174CC
Jorvet 16g x 1in. Hypodermic needle is made of stainless steel, has a round hub, and is packaged in a cardboard box or clear plastic case. This product contains 12 needles.